2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromospiro[fluorene-9,9'-xanthene]

B5842-1G B5842-5G
TCB5842-1GEA 70 USD
TCB5842-1G TCB5842-5G
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)
2-Bromospiro[9H-fluorene-9,9'-[9H]xanthene]
Formula: C₂₅H₁₅BrO
MDL Number: MFCD29050113
CAS Number: 899422-06-1

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR