trans,trans-4'-Butylbicyclohexyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: trans,trans-4'-Butyl-1,1'-bicyclohexane-4-carboxylic acid, trans-4-(trans-4-Butylcyclohexyl)cyclohexanecarboxylic acid

B5317-1G B5317-5G
TCB5317-1GEA 88 USD
TCB5317-1G TCB5317-5G
trans,trans-4'-Butylbicyclohexyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
trans,trans-4'-Butylbicyclohexyl-4-carboxylic acid
Formula: C₁₇H₃₀O₂
MW: 266.43 g/mol
MDL Number: MFCD06797517
CAS Number: 89111-63-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Mesomorphic range 211 to 243 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR