2-Bromo-9,9-dibutylfluorene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5127-1G B5127-5G
TCB5127-1GEA 77 USD
TCB5127-1G TCB5127-5G
2-Bromo-9,9-dibutylfluorene ≥98.0% (by GC)
2-Bromo-9,9-dibutylfluorene
Formula: C₂₁H₂₅Br
MDL Number: MFCD12031333
CAS Number: 88223-35-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR