3-Butyn-1-amine hydrochloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

B5925-1G B5925-200MG
TCB5925-1GEA 162 USD
TCB5925-1G TCB5925-200MG
3-Butyn-1-amine hydrochloride ≥95.0% (by titrimetric analysis)
3-Butyn-1-amine hydrochloride
Formula: C₄H₇N·HCl
MDL Number: MFCD06658391
CAS Number: 88211-50-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR