[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-4-[(tert-butyldimethylsilyloxy)methyl]benzene

B5155-25G B5155-5G
TCB5155-25GEA 368 USD
TCB5155-25G TCB5155-5G
[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane ≥98.0% (by GC)
[(4-Bromobenzyl)oxy](tert-butyl)dimethylsilane
Formula: C₁₃H₂₁BrOSi
MW: 301.30 g/mol
Boiling Pt: 160 °C
MDL Number: MFCD07368899
CAS Number: 87736-74-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR