(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: (E,E)-1,4-Bis[2-[4-[N,N-bis(4-methoxyphenyl)amino]phenyl]vinyl]benzene, (E,E)-4,4'-[1,4-Phenylenebis(ethene-2,1-diyl)]bis[N,N-bis(4-methoxyphenyl)aniline], TOP-HTM-α1

B5672-1G B5672-25G B5672-5G
TCB5672-1GEA 199 USD
TCB5672-1G TCB5672-25G TCB5672-5G
(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene ≥98.0% (by HPLC, total nitrogen)
(E,E)-1,4-Bis[4-[bis(4-methoxyphenyl)amino]styryl]benzene
Formula: C₅₀H₄₄N₂O₄
MW: 736.91 g/mol
Melting Pt: 181 °C
CAS Number: 872466-50-7

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR