1,6-Dibromo-3,8-diisopropylpyrene ≥98.0% (by GC)

Supplier: TCI America

D5236-1G D5236-200MG
TCD5236-1GEA 443 USD
TCD5236-1G TCD5236-200MG
1,6-Dibromo-3,8-diisopropylpyrene ≥98.0% (by GC)
1,6-Dibromo-3,8-diisopropylpyrene
Formula: C₂₂H₂₀Br₂
MDL Number: MFCD27979119
CAS Number: 869340-02-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR