4-Cyano-3-fluorophenyl 4-Propylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Cyano-3-fluorophenyl 4-propylbenzoate, 4-Propylbenzoic acid 4-Cyano-3-fluorophenyl ester

C3554-1G C3554-5G
TCC3554-1GEA 30 USD
TCC3554-1G TCC3554-5G
4-Cyano-3-fluorophenyl 4-Propylbenzoate ≥98.0% (by GC)
4-Cyano-3-fluorophenyl 4-Propylbenzoate
Formula: C₁₇H₁₄FNO₂
MW: 283.30 g/mol
MDL Number: MFCD06797522
CAS Number: 86776-51-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR