Belinostat ≥98.0% (by HPLC)

Supplier: TCI America

B5888-100MG B5888-25MG
TCB5888-100MGEA 177 USD
TCB5888-100MG TCB5888-25MG
Belinostat ≥98.0% (by HPLC)
Belinostat
Formula: C₁₅H₁₄N₂O₄S
MDL Number: MFCD08064035
CAS Number: 866323-14-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR