4,4'-Bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,4'-Bis(trans-4-propylcyclohexyl)-1,1'-biphenyl

B5881-25G B5881-5G
TCB5881-25GEA 201 USD
TCB5881-25G TCB5881-5G
4,4'-Bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
4,4'-Bis(trans-4-propylcyclohexyl)biphenyl
Formula: C₃₀H₄₂
MW: 402.67 g/mol
MDL Number: MFCD11053445
CAS Number: 85600-56-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR