5-(Bromomethyl)undecane ≥97.0% (by GC)

Supplier: TCI America
Warning

B5721-25G B5721-5G
TCB5721-25GEA 490 USD
TCB5721-25G TCB5721-5G
5-(Bromomethyl)undecane ≥97.0% (by GC)
5-(Bromomethyl)undecane
Formula: C₁₂H₂₅Br
MW: 249.24 g/mol
Boiling Pt: 85 °C
MDL Number: MFCD23115881
CAS Number: 85531-02-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR