3-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5869-1G B5869-5G
TCB5869-1GEA 109 USD
TCB5869-1G TCB5869-5G
3-(3-Bromophenyl)-9-phenyl-9H-carbazole ≥98.0% (by GC)
3-(3-Bromophenyl)-9-phenyl-9H-carbazole
Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
MDL Number: MFCD32062864
CAS Number: 854952-59-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 179.0 to 183.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR