3,3'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,3'-Biphenyldiboronic acid bis(pinacol) ester

B5704-1G B5704-200MG
TCB5704-1GEA 339 USD
TCB5704-1G TCB5704-200MG
3,3'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl ≥98.0% (by GC, titration analysis)
3,3'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl
Formula: C₂₄H₃₂B₂O₄
MW: 406.14 g/mol
CAS Number: 850264-92-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR