2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromonaphthalene-1,4,5,8-tetracarboxylic Dianhydride, 4-Bromoisochromeno[6,5,4-def]isochromene-1,3,6,8-tetraone
B5756-1G
B5756-5G
TCB5756-1GEA
236
USD
TCB5756-1G
TCB5756-5G
2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride ≥98.0% (by titrimetric analysis)
2-Bromonaphthalene-1,4,5,8-tetracarboxylic 1,8:4,5-Dianhydride
Formula:
C₁₄H₃BrO₆ |
CAS Number:
846-20-8 |
Specification Test Results
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...