3-Amino-2-chloro-6-methylphenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-4-methyl-3-hydroxyaniline, 5-Amino-6-chloro-o-cresol

A3266-25G A3266-5G
TCA3266-25GEA 203 USD
TCA3266-25G TCA3266-5G
3-Amino-2-chloro-6-methylphenol ≥98.0% (by GC, titration analysis)
3-Amino-2-chloro-6-methylphenol
Formula: C₇H₈ClNO
MW: 157.60 g/mol
MDL Number: MFCD03094650
CAS Number: 84540-50-1
EINECS: 283-144-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR