9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene ≥98.0% (by GC)
Supplier: TCI America
B5718-1G
B5718-5G
TCB5718-1GEA
86
USD
TCB5718-1G
TCB5718-5G
9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene ≥98.0% (by GC)
9-([1,1'-Biphenyl]-3-yl)-10-bromoanthracene
Formula:
C₂₆H₁₇Br MW: 409.33 g/mol |
MDL Number:
MFCD30470615 CAS Number: 844679-02-3 |
Specification Test Results
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...