alpha-Arbutin ≥95.0% (by HPLC)

Supplier: TCI America

A3284-25G A3284-5G
TCA3284-25GEA 92 USD
TCA3284-25G TCA3284-5G
alpha-Arbutin ≥95.0% (by HPLC)
alpha-Arbutin
Formula: C₁₂H₁₆O₇
MDL Number: MFCD09838262
CAS Number: 84380-01-8
EINECS: 440-470-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 204.0 to 208.0 deg-C
Specific rotation [a]20/D +174 to +186 deg (C=1, water)
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR