18-(tert-Butoxy)-18-oxooctadecanoic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: tert-Butyl Hydrogen Octadecanedioate, Mono-tert-butyl Octadecanedioate, tBu ester-C16-acid, Octadecanedioic acid Mono-tert-butyl ester

B5928-1G B5928-250MG
TCB5928-1GEA 262 USD
TCB5928-1G TCB5928-250MG
18-(tert-Butoxy)-18-oxooctadecanoic acid ≥95.0% (by GC, titration analysis)
18-(tert-Butoxy)-18-oxooctadecanoic acid
Formula: C₂₂H₄₂O₄
MDL Number: MFCD09991735
CAS Number: 843666-40-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR