tert-Butyl Hydrogen Hexadecanedioate ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 16-(tert-Butoxy)-16-oxohexadecanoic acid, Hexadecanedioic acid Mono-tert-butyl ester, Boc-C14-COOH

B4911-100MG B4911-1G
TCB4911-100MGEA 102 USD
TCB4911-100MG TCB4911-1G
tert-Butyl Hydrogen Hexadecanedioate ≥95.0% (by GC, titration analysis)
tert-Butyl Hydrogen Hexadecanedioate
Formula: C₂₀H₃₈O₄
MDL Number: MFCD23136039
CAS Number: 843666-27-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR