9-(3-Bromopropyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5183-1G B5183-200MG
TCB5183-1GEA 325 USD
TCB5183-1G TCB5183-200MG
9-(3-Bromopropyl)carbazole ≥98.0% (by GC)
9-(3-Bromopropyl)carbazole
Formula: C₁₅H₁₄BrN
MDL Number: MFCD11164489
CAS Number: 84359-61-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR