4,4''-Dibromo-1,1':3',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,4''-Dibromo-m-terphenyl

D5023-1G D5023-200MG
TCD5023-1GEA 266 USD
TCD5023-1G TCD5023-200MG
4,4''-Dibromo-1,1':3',1''-terphenyl ≥98.0% (by GC)
4,4''-Dibromo-1,1':3',1''-terphenyl
Formula: C₁₈H₁₂Br₂
MDL Number: MFCD28138091
CAS Number: 83909-22-2
UN: 3152
ADR: 9,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR