4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)

Supplier: TCI America
Danger

C3329-1G C3329-200MG
TCC3329-1GEA 289 USD
TCC3329-1G TCC3329-200MG
4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)
4-(Chloromethyl)-4'-methyl-2,2'-bipyridyl
Formula: C₁₂H₁₁ClN₂
MDL Number: MFCD24464190
CAS Number: 83799-54-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR