SteriWare Multi Grip Tweezers
Supplier: SAMPLING SYSTEMS LTD
8270A-200 8270A-200S 8270A-300 8270A-300S
77577-230EA 133.36 USD
77577-230 77577-232 77577-234 77577-236
SteriWare Multi Grip Tweezers
Forceps
These SteriWare multi grip tweezers operate like conventional tweezers, they have an additional grip feature enabling the easy and secure handling of small vials and containers.
  • Made from food grade PS
  • Individually bagged to prevent cross contamination
  • 100% manufactured and packed in a cleanroom
  • Choice of sterile and non sterile