SteriWare Micro-Spatula Samplers
Supplier: SAMPLING SYSTEMS LTD
8015NB-170S 8015R-170S 8015VB-170S 8015B-170S
77576-654EA 142.42 USD
77576-654 77576-658 77576-664 77576-644
SteriWare Micro-Spatula Samplers
Samplers Solid Samplers
Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.
  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use
Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.