Slot Samplers
Supplier: SAMPLING SYSTEMS LTD
1210A-1000 1210A-1500 1210A-2000 1210A-300 1210A-600 1210B-1000 1210K-1000 1210K-1500 1210K-2000
77576-042EA 804.38 USD
77576-042 77576-044 77576-046 77576-048 77576-050 77576-052 77576-056 77576-058 77576-060
Slot Samplers
Samplers Solid Samplers
These all layer samplers have a robust all stainless steel construction and are suitable for sampling free flowing powders, granules and even slightly cohesive powders.
  • Multilevel samplers
  • Cross section type
  • With or without bottle holder
For taking a large volume, cross sectional sample from a container.
Información para pedidos: Sample collecting bottles (for use with the sampler with bottle holder) as well as cleaning brushes can be ordered separately.