Cup Samplers
Supplier: SAMPLING SYSTEMS LTD
5030A-1000 5030A-1500 5030A-600 5030J-1000 5030J-1500
77576-422EA 478.1 USD
77576-422 77576-424 77576-426 77576-428 77576-430
Cup Samplers
Samplers Fluid Samplers
Stainless steel cup samplers designed for liquids and suspensions.
  • Single point liquid sampler with removable cup
  • Simple operation, easy to clean and hygienic design
  • Discrete samples to be taken from known heights within the product