SteriWare® ViscoThief Disposable Samplers
Supplier: SAMPLING SYSTEMS LTD
8055P-500 8055H-1000 8055H-1000S 8055H-1500 8055H-1500S 8055H-500 8055H-500S 8055P-1000
77576-906EA 219.64 USD
77576-906 77576-892 77576-894 77576-896 77576-898 77576-900 77576-902 77576-904
SteriWare® ViscoThief Disposable Samplers
Samplers Disposable Samplers
Disposable viscous sampler is designed for creams, pastes and gels. The robust HDPE and PP construction allows for a huge number of liquid and low viscosity testing applications.
  • No risk of cross-contamination (device can be used straight from the bag)
  • Silicone seal for improved sampling performance
  • Assembled and packed in a class 100000 cleanroom
Suitable for taking fast samples from tanks, kegs and containers in industries such as pharmaceutical, food, chemical, cosmetic, biotechnology. Pre-sterilised versions are available.
Certificaciones: FDA, EU 10/2011 and EC 1935/2004 conformity. BSE/TSE free. Full batch traceability, ATEX rated.
Envase: Individually bagged in a heat-sealed PE bag.