Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Supplier: Thermo Fisher Scientific
B1A-8 B1A-6 B2-16 B2-24 B2-WALL B2-RL-9D B2-RL-25D B2-RL-25C B2-RL-9C B1-RL-9D B1-RL-9C B1-RL-17D B1-RL-17C B1A-5C B1A-5D B1A-PREP
14005-882EA 104.77 USD
14005-882 14005-884 14005-886 14005-888 14005-890 14005-892 14005-894 14005-896 14005-898 14006-030 14007-028 27372-214 27372-216 27372-222 27372-224 27372-288 27373-254 27373-272 27373-274 27373-276 27373-278 27373-280 27373-282 27373-284 27373-312 27373-313 27373-314 27373-315 27373-316 27373-317 27373-318 27372-290
Combs for Owl™ EasyCast™ Mini Gel Electrophoresis System, Models B1, B2 and B3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Use comb with your exisitng U.V
  • For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems
  • Cast smaller gels
For use in Owl™ EasyCast™ Mini Gel Electrophoresis Systems. Use comb with your exisitng U.V. Transmissible (UVT) gel tray and comb slots to cast smaller gels. Thickness: 1.5mm (1/16").
Transmissible (UVT) gel tray and comb slots to cast smaller gels.