MaxiCut® Ultra™ 44-5745E Seamless Knit Gloves with Nitrile Coating
Supplier: PIP
44-5745E/XS 44-5745E/XXL 44-5745E/L 44-5745E/M 44-5745E/S 44-5745E/XL
77531-192CS 1115.45 USD
77531-192 77531-184 77531-186 77531-210 77531-194 77531-190
MaxiCut® Ultra™ 44-5745E Seamless Knit Gloves with Nitrile Coating
Gloves
Designed and developed as a 360° breathable cut resistant glove, MaxiCut® Ultra™ combines high levels of cut protection with comfort, flexibility and dexterity for dry environments.
  • Innovative fibers ATG sources their base ingredients to develop their own unique high performance yarns and fibers that offer cut protection while maintaining high levels of comfort
  • 360° breathability patented Nitrile MicroFoam coating offers 360° breathability
  • Reinforcement between the thumb and forefinger, improves cut resistance and extends glove life in an inherently weak area
  • Consistent liner penetration from the nitrile, leaving only the soft liner resting against the skin
  • Form, fit and feel mimics the 'hand at rest', reducing hand fatigue and increasing comfort
  • Touchscreen compatible to allow the user to operate a touch screen phone or device without removing gloves
  • Latest knitting technology utilized to produce smooth and rounded fingertips, improving fingertip sensitivity
  • 100% silicone free
All ingredients used in the manufacturing and construction of this product are compliant with the REACH legislation.
The Oeko-Tex Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin. The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product.
This glove has been laundered prior to final packaging making it clean and safe for use.
Certificaciones: ANSI A5 EN3.