3-Chloro-5-fluoroisonicotinic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-5-fluoro-4-pyridinecarboxylic acid

A999964-1G A999964-250MG
77873-410EA 103 USD
77873-410 77873-411
3-Chloro-5-fluoroisonicotinic acid ≥98%
3-Chloro-5-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD13185823
CAS Number: 514798-03-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR