(2,6-Dimethoxypyridin-4-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A999484-250MG A999484-1G A999484-100MG
77784-767EA 328 USD
77784-767 77784-766 77784-765
(2,6-Dimethoxypyridin-4-yl)boronic acid ≥98%
(2,6-Dimethoxypyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
MDL Number: MFCD18072610
CAS Number: 1220188-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR