4-Vinyl-dihydrofuran-2(3H)-one ≥97% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with TBC

A998436-1G A998436-250MG A998436-5G
77595-612EA 110.7 USD
77595-612 77618-720 77749-838
4-Vinyl-dihydrofuran-2(3H)-one ≥97% stabilized
4-Vinyl-dihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₆H₈O₂
MDL Number: MFCD13181922
CAS Number: 53627-36-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR