(4-Methanesulfonyl-2-methylphenyl)boronic acid ≥97%

Supplier: Ambeed
Warning

A997167-5G A997167-250MG A997167-1G
77749-835EA 607 USD
77749-835 77749-834 77749-833
(4-Methanesulfonyl-2-methylphenyl)boronic acid ≥97%
(4-Methanesulfonyl-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄S
MDL Number: MFCD27935675
CAS Number: 850033-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR