Uridine 5'-diphosphate sodium salt ≥98%

Supplier: Ambeed
Danger

A996472-1G A996472-250MG A996472-5G A996472-100MG
77599-452EA 59.45 USD
77599-452 77599-454 77599-456 77623-450
Uridine 5'-diphosphate sodium salt ≥98%
Uridine 5'-diphosphate sodium salt

Precaución: for Research Use Only

Formula: C₉H₁₄N₂NaO₁₂P₂
MDL Number: MFCD30528029
CAS Number: 21931-53-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR