3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(3-nitro-4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane

A996124-5G A996124-1G A996124-250MG A996124-100MG
77784-741EA 999 USD
77784-741 77784-739 77784-740 77784-738
3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester ≥97%
3-nitro-4-(trifluoromethoxy)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₃NO₅
MDL Number: MFCD21333035
CAS Number: 2096336-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR