Potassium (4-(9H-carbazol-9-yl)phenyl)trifluoroborate ≥95%

Supplier: Ambeed
Warning

A995939-1G A995939-100MG A995939-5G A995939-250MG
77623-440EA 99.91 USD
77623-440 77784-733 77784-735 77784-734
Potassium (4-(9H-carbazol-9-yl)phenyl)trifluoroborate ≥95%
Potassium (4-(9H-carbazol-9-yl)phenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₈H₁₂BF₃KN
MDL Number: MFCD20486766
CAS Number: 1874177-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR