3,5-Dimethyl-1H-pyrazole-1-carbothioamide ≥85%

Supplier: Ambeed
Warning

A995887-5G A995887-1G A995887-250MG A995887-100MG
77599-450EA 867.15 USD
77599-450 77784-731 77784-732 77784-730
3,5-Dimethyl-1H-pyrazole-1-carbothioamide ≥85%
3,5-Dimethyl-1H-pyrazole-1-carbothioamide

Precaución: for Research Use Only

Formula: C₆H₉N₃S
MDL Number: MFCD00180878
CAS Number: 1124-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR