N'-Hydroxy-2,2-dimethylpropanimidamide ≥95%

Supplier: Ambeed
Warning

A995588-1G A995588-5G
77573-986EA 42 USD
77573-986 77784-726
N'-Hydroxy-2,2-dimethylpropanimidamide ≥95%
N'-Hydroxy-2,2-dimethylpropanimidamide

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O
MDL Number: MFCD03426250
CAS Number: 42956-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR