tert-Butyl 2-oxocyclopentane-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A995577-1G A995577-5G
77784-724EA 334 USD
77784-724 77784-725
tert-Butyl 2-oxocyclopentane-1-carboxylate ≥98%
tert-Butyl 2-oxocyclopentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₃
MDL Number: MFCD19301646
CAS Number: 84109-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR