5,7-Dimethoxy-8-(1-(4-methoxyphenyl)-3-oxo-3-(pyrrolidin-1-yl)propyl)-4-phenyl-2h-chromen-2-one ≥98%

Supplier: Ambeed
Warning

Synonyms: CMLD-2

A995039-10MG A995039-1MG A995039-5MG
77599-444EA 99.43 USD
77599-444 77599-446 77599-448
5,7-Dimethoxy-8-(1-(4-methoxyphenyl)-3-oxo-3-(pyrrolidin-1-yl)propyl)-4-phenyl-2h-chromen-2-one ≥98%
5,7-Dimethoxy-8-(1-(4-methoxyphenyl)-3-oxo-3-(pyrrolidin-1-yl)propyl)-4-phenyl-2h-chromen-2-one

Precaución: for Research Use Only

Formula: C₃₁H₃₁NO₆
MW: 513.58 g/mol
MDL Number: MFCD32197047
CAS Number: 958843-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR