(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-methyl-1H-indol-3-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp(6-Me)-OH

A995010-1G A995010-250MG
77784-717EA 493 USD
77784-717 77784-718
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-methyl-1H-indol-3-yl)propanoic acid ≥95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-methyl-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MW: 440.4905 g/mol
MDL Number: MFCD29917531
CAS Number: 908846-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR