(2aS,4S,5'R,6aS,6bS,8aS,8bR,9S,10R,11aS,12aS,12bR)-5',6a,8a,9-Tetramethyldocosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: Tigogenin

A993709-5G A993709-10MG
77687-012EA 9 USD
77687-012 77687-011
(2aS,4S,5'R,6aS,6bS,8aS,8bR,9S,10R,11aS,12aS,12bR)-5',6a,8a,9-Tetramethyldocosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol ≥98%
(2aS,4S,5'R,6aS,6bS,8aS,8bR,9S,10R,11aS,12aS,12bR)-5',6a,8a,9-Tetramethyldocosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol

Precaución: for Research Use Only

Formula: C₂₇H₄₄O₃
MW: 416.64 g/mol
MDL Number: MFCD00067287
CAS Number: 77-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR