1-(3-Chloro-4-methylphenyl)ethan-1-amine ≥95%

Supplier: Ambeed
Danger

A993091-100MG A993091-250MG
77623-424EA 65.92 USD
77623-424 77623-426
1-(3-Chloro-4-methylphenyl)ethan-1-amine ≥95%
1-(3-Chloro-4-methylphenyl)ethan-1-amine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN
MW: 169.65 g/mol
MDL Number: MFCD18702551
CAS Number: 105321-42-4
UN: 2735
ADR: 8 Ⅲ

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR