Fmoc-Ala-Cl ≥95%

Supplier: Ambeed
Danger

A993011-1G A993011-250MG A993011-5G
77595-606EA 124.03 USD
77595-606 77880-809 77880-810
Fmoc-Ala-Cl ≥95%
Fmoc-Ala-Cl

Precaución: for Research Use Only

Formula: C₁₈H₁₆NO₃Cl
MDL Number: MFCD00235802
CAS Number: 103321-50-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR