3,6,9,12,15,18,21,24,27,30,33,36,39,42,45-Pentadecaoxaheptatetracontane-1,47-diol ≥95%

Supplier: Ambeed
Warning

Synonyms: HO-PEG16-OH

A991931-100MG A991931-250MG
77599-430EA 192.7 USD
77599-430 77599-432
3,6,9,12,15,18,21,24,27,30,33,36,39,42,45-Pentadecaoxaheptatetracontane-1,47-diol ≥95%
3,6,9,12,15,18,21,24,27,30,33,36,39,42,45-Pentadecaoxaheptatetracontane-1,47-diol

Precaución: for Research Use Only

Formula: C₃₂H₆₆O₁₇
MW: 722.86 g/mol
MDL Number: MFCD09910395
CAS Number: 6812-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR