(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(4-chlorophenyl)propanoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-Me-Phe(4-Cl)-OH

A991234-5G A991234-100MG A991234-250MG A991234-1G
77573-974EA 115 USD
77573-974 77784-670 77784-672 77784-671
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(4-chlorophenyl)propanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(4-chlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₂ClNO₄
MDL Number: MFCD04974251
CAS Number: 1217716-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR