3-(4-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥95%

Supplier: Ambeed
Warning

A991227-1G A991227-100MG A991227-250MG A991227-5G
77595-600EA 246 USD
77595-600 77599-418 77599-420 77623-402
3-(4-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione ≥95%
3-(4-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁FN₂O₃
MDL Number: MFCD31556181
CAS Number: 2359705-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR