4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide ≥98%

Supplier: Ambeed
Warning

A991078-1MG A991078-5MG A991078-10MG
77599-414EA 28.7 USD
77599-414 77599-416 77623-400
4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide ≥98%
4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₆H₁₉F₃N₄O₂
CAS Number: 1204329-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR