(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A990911-100MG A990911-1G
77886-958EA 72 USD
77886-958 77886-959
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid ≥95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₅
MDL Number: MFCD04117844
CAS Number: 1217662-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR