tert-Butyl (2-amino-2-methylpropyl)carbamate ≥98%

Supplier: Ambeed
Warning

A990829-5G A990829-1G A990829-250MG
77573-972EA 312 USD
77573-972 77886-956 77886-957
tert-Butyl (2-amino-2-methylpropyl)carbamate ≥98%
tert-Butyl (2-amino-2-methylpropyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₂
MDL Number: MFCD11040212
CAS Number: 95034-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR